Learn More
AG Scientific Inc Chymostatin, microbial, 25 MG
Supplier:  AG Scientific Inc C146925MG
Encompass_Preferred
Chymostatin is a reversible serine and cysteine proteases inhibitor, blocking strongly chymotrypsin (ID=150g/ml) and chymase and weakly papin(ID= 7.5g/ml), cathepsins and leukocyte elastase. It also reduces myocardial active TGF-β levels but it does not affect trypsin, thrombin, plasmin, pepsin and kallikrein.CAS Number: 9076-44-2Molecular Weight: 604.24 DaChemical Formula: C31H41N7O6Solubility: Up to 20 mg/mL in DMSO, up to 10 mg/mL in 5% Glacial Acetic AcidStorage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7011