Learn More
Selleck Chemical LLC Canertinib (CI-1033) 10mg 267243-28-7 PD183805
SDP
Supplier:  Selleck Chemical LLC S101910MG
Encompass_Preferred
Canertinib (CI-1033, PD183805) is a pan-ErbB inhibitor for EGFR and ErbB2 with IC50 of 1.5 nM and 9.0 nM, no activity to PDGFR, FGFR, InsR, PKC, or CDK1/2/4. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-862-9