Learn More
Selleck Chemical LLC PD184352 (CI-1040) 5mg 212631-79-3
SDP
Supplier:  Selleck Chemical LLC S10205MG
Encompass_Preferred
PD184352 (CI-1040) is an ATP non-competitive MEK1/2 inhibitor with IC50 of 17 nM in cell-based assays, 100-fold more selective for MEK1/2 than MEK5. PD184352 (CI-1040) selectively induces apoptosis. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-871-5