Learn More
Medchemexpress LLC HY-100246 5mg Medchemexpress, CI-1044 CAS:197894-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1002465MG
Encompass_Preferred
Medchemexpress, HY-100246 5mg CI-1044 CAS:197894-84-1 CI-1044 is an orally active PDE4 inhibitor with IC50s of 0.29, 0.08, 0.56, 0.09 μM for PDE4A5, PDE4B2, PDE4C2 and PDE4D3, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0581