Learn More
Medchemexpress LLC CI-966 hydrochloride | 110283-66-4 | 509.87 g/mol | C23H22ClF6NO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10353410MM 1ML
Encompass_Preferred
CI-966 hydrochloride is a selective inhibitor of the GABA transporter GAT-1 used as a research reagent for in vitro and in vivo studies of GABAergic signaling. It is the hydrochloride salt and has reported potency at human and rat GAT-1.
  • Selective inhibitor of GAT-1 with reported IC50s of 0.26 μM (hGAT-1) and 1.2 μM (rGAT-1).
  • Orally active and brain-penetrant, useful for neuropharmacology studies.
  • Soluble in DMSO; clear solutions at reported concentrations.
  • Typical handling: solid stored at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (short term).
Catalog No. 50-002-46167