Learn More
Medchemexpress LLC cIAP1 ligand 1 | 2095244-42-9 | >99.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128808100MG
Encompass_Preferred
cIAP1 ligand 1 is an LCL161 derivative based IAP ligand that can be used in prostate cancer research. It can be connected to the ABL ligand for protein by a linker to form SNIPER. This product is a PROTAC, which are heterobifunctional nanomolecules that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • LCL161 derivative based IAP ligand
  • Can be connected to ABL ligand for protein by a linker to form SNIPER
  • Useful in prostate cancer research
  • Exploits intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-38980