Learn More
Medchemexpress LLC Cibenzoline 100mg | 53267-01-9 | 100MG
SDP
Supplier:  Medchemexpress LLC HY106577100MG
Encompass_Preferred
Cibenzoline is a class Ia antiarrhythmic active molecule with low anticholinergic activity Cibenzoline is a KATP channel inhibitor acting through the pore forming subunit Kir6 2 with an IC50 of 22 2 M Cibenzoline inhibits IKr and IKs currents with IC50 values of 8 8 M and 12 3 M respectively Cibenzoline is used in the study of cardiac diseases In addition Cibenzoline can induce hypoglycemia[1 [2 [3 [4
Catalog No. 50-002-41028