Learn More
Medchemexpress LLC Cibenzoline | 53267-01-9 | MFCD00864706 | ≥95.0% | 262.35 g/mol | C18H18N2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10657750MG
Encompass_Preferred
Cibenzoline is provided as an analytical standard for laboratory research. It is a class Ia antiarrhythmic (cifenline) and KATP channel inhibitor supplied as a solid reference material for qualitative and quantitative analysis, method development, and validation in chromatography and mass spectrometry.
  • Intended for research use and analytical reference.
  • Suitable for HPLC, GC, and MS method development and validation.
  • High purity: ≥95.0% (analytical standard grade).
  • Formula C18H18N2; molecular weight 262.35 g/mol.
  • Provided as a solid powder in defined pack sizes for precise dosing.
Catalog No. 50-002-40963