Learn More
Medchemexpress LLC Cibenzoline | 53267-01-9 | MFCD00864706 | 95.1% | 262.35 | C18H18N2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10657710MG
Encompass_Preferred
Cibenzoline is an analytical reference standard of a class Ia antiarrhythmic imidazoline compound intended for research and analytical applications. It is provided for use in qualitative and quantitative assays and method development for chromatographic and mass spectrometric workflows.
  • Analytical reference standard for HPLC, GC, and MS assays.
  • Suitable for qualitative and quantitative method development.
  • Used in pharmacology and ion channel research applications.
  • Molecular formula C18H18N2; molecular weight 262.35.
  • Reported purity approximately 95.1% (manufacturer documentation).
Catalog No. 50-002-40448