Learn More
Medchemexpress LLC Ciclesonide | 126544-47-6 | MFCD00866004 | 100.0% | 540.7 g/mol | C32H44O7 | 10 MG

Supplier: Medchemexpress LLC HYB0625R10MG
Ciclesonide analytical standard supplied as a 10 mg reference material for analytical and research use. It is the analytical standard of the glucocorticoid ciclesonide commonly used in HPLC, GC, and MS assays. Purity is listed at 99.98% (molecular formula C32H44O7; CAS 126544-47-6).
- High purity suitable for analytical applications.
- Optimized for HPLC, GC, and MS methods.
- Convenient 10 mg quantity for reference standard use.
- Molecular weight 540.7 g/mol for accurate calculations.
- CAS number 126544-47-6 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.