Learn More
Apexbio Technology LLC Ciclopirox 29342-05-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20875.1
Encompass_Preferred
Ciclopirox is a synthetic antifungal compound with broad-spectrum activity against dermatophytes yeasts and molds It disrupts fungal membranes and interferes with essential enzymes inhibiting cell growth and replication Ciclopirox also has antibacterial properties against select Gram-positive and Gram-negative bacteria In vitro it demonstrates anti-inflammatory and anticancer activities suggesting its potential in research on fungal infections inflammation and tumorigenesis The compound s inhibitory concentrations (IC50) in antifungal assays typically range from 0 5 to 5 g/mL depending on the target species and conditions Ciclopirox is used to study fungal pathogenicity and develop therapies for dermatopharmacology and oncology
Catalog No. 50-000-61804