Learn More
Medchemexpress LLC Ciclopirox | 29342-05-0 | 98.3% | 1 ML

Supplier: Medchemexpress LLC HYB045010MM/1ML
Ciclopirox is a synthetic, orally active antifungal agent used in research for superficial mycoses. It demonstrates broad-spectrum activity against dermatophytes, yeasts, molds, and various pathogenic bacteria, and also exhibits anticancer and anti-inflammatory effects.
- Synthetic, orally active antifungal agent
- Used for superficial mycoses research
- Exhibits broad-spectrum activity against dermatophytes, yeasts, molds, and pathogenic bacteria
- Demonstrates anticancer and anti-inflammatory effects
- Available as a 10 mM x 1 mL solution in DMSO
- Shipped at room temperature
- Powder storage: -20°C for 3 years or 4°C for 2 years
- In-solvent storage: -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.