Learn More
Selleck Chemical LLC Ciclopirox 200mg 29342-05-0
SDP
Supplier:  Selleck Chemical LLC S2528200MG
Encompass_Preferred
Ciclopirox is a broad-spectrum antifungal agent working as an iron chelator. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-648-1