Learn More
Selleck Chemical LLC Ciclopirox ethanolamine S3019-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S301910MM/1ML
Encompass_Preferred
Ciclopirox ethanolamine (Ciclopirox olamine HOE 296 Ciclopiroxolamine) is a broad-spectrum antifungal agent working as an iron chelator
Catalog No. 50-311-1602