Learn More
Medchemexpress LLC Ciclopirox olamine | 41621-49-2 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0450A10MM/1ML
Encompass_Preferred
Ciclopirox olamine is a synthetic and orally active antifungal agent used for superficial mycoses research. It has a very broad spectrum of activity, inhibiting dermatophytes, yeasts, molds, and many pathogenic Gram-positive and Gram-negative species. Additionally, Ciclopirox olamine exhibits anticancer and anti-inflammatory effects.
  • Synthetic and orally active antifungal agent
  • Broad spectrum of activity against dermatophytes, yeasts, molds, and many pathogenic Gram-positive and Gram-negative species
  • Exhibits anticancer effects
  • Exhibits anti-inflammatory effects
Catalog No. 50-004-05448