Learn More
Cayman Chemical CIcloprofen 50mg
SDP
Supplier:  Cayman Chemical 3357150 MG
Encompass_Preferred
An NSAID; inhibits ragweed-, A23187-, or concanavalin A-induced histamine release in isolated human leukocytes at 0.5 mM; inhibits carrageenan-induced paw edema in rats (ED50 = 67 mg/kg)
Catalog No. 50-294-6153