Learn More
Medchemexpress LLC Cid 16020046 | 834903-43-4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16697
Encompass_Preferred
CID 16020046 is a potent and selective GPR55 antagonist that inhibits GPR55 constitutive activity with an IC50 of 0.15 μM. It also inhibits GPR55-mediated Ca2+ signaling and ERK1/2 phosphorylation. Furthermore, CID 16020046 reduces wound healing in endothelial cells and is involved in the regulation of platelet function.
  • Potent and selective GPR55 antagonist
  • Inhibits GPR55 constitutive activity (IC50 of 0.15 μM)
  • Inhibits GPR55-mediated Ca2+ signaling
  • Inhibits GPR55-mediated ERK1/2 phosphorylation
  • Reduces wound healing in endothelial cells
  • Involved in the regulation of platelet function
Catalog No. 50-003-92576