Learn More
Cayman Chemical CID-2745687 10mg
SDP
Supplier:  Cayman Chemical 1204610 MG
Encompass_Preferred
A reversible, competitive antagonist of GPR35, blocking activation by the synthetic agonist pamoic acid with a Ki of 12.8 nM; less potently blocks activation of GPR35 by zaprinast (IC50 = 160 nM); shows ~57-fold selectivity for GPR35 over the related receptor GPR55 (IC50 = 9.08 µM)
Catalog No. 50-295-3763