Learn More
Cayman Chemical CID-5951923 10mg
SDP
Supplier:  Cayman Chemical 2164910 MG
Encompass_Preferred
A KLF5 inhibitor (IC50 = 2.3 μM); decreases KLF5 expression in and reduces growth of DLD-1 colorectal adenocarcinoma cells in a concentration-dependent manner; selectively inhibits proliferation of cancer cell lines that highly express KLF5 in the NCI-60 panel of cancer cell lines
Catalog No. 50-298-6063