Learn More
Medchemexpress LLC CID 697851 | 312508-42-2 | C14H12N4OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1512215MG
Encompass_Preferred
CID 697851 is a solid, light yellow to yellow compound that functions as an AddAB and RecBCD inhibitor. It offers IC50s of 13 and 33 nM, making it suitable for antibacterial research.
  • Functions as an AddAB and RecBCD inhibitor
  • Offers IC50s of 13 and 33 nM
  • Suitable for antibacterial research
  • High purity (99.49%)
  • Solid appearance
  • Color ranges from light yellow to yellow
Catalog No. 50-003-75820