Learn More
TARGETMOL CHEMICALS INC CID755673 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T245825MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CID755673 is an effective and specific cell-active inhibitor for PKD (IC50 182 nM); exhibits selective PKD1 inhibition when compared with PLK1 AKT CAMKII(alpha) CAK and PKC isoforms. purity: 99%
Catalog No. 50-257-7117