Learn More
Medchemexpress LLC Cidofovir | 113852-37-2 | 99.95% | 279.19 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1743850MG
Encompass_Preferred
Cidofovir is an acyclic monophosphate nucleotide analogue and a potent inhibitor of cytomegalovirus (CMV). It exerts its antiviral activity by selectively inhibiting viral DNA polymerase. This compound induces apoptosis and is utilized in studies concerning AIDS cytomegalovirus retinitis, herpes, and various cancers. Additionally, it demonstrates anti-orthopoxvirus and anti-variola activities.
  • Acyclic monophosphate nucleotide analogue
  • Inhibits cytomegalovirus (CMV) replication
  • Selectively inhibits viral DNA polymerase
  • Induces apoptosis
  • Used in studies of AIDS cytomegalovirus retinitis, herpes, and cancer
  • Exhibits anti-orthopoxvirus and anti-variola activities
  • High purity of 99.95%
Catalog No. 50-003-86632