Learn More
Enzo Life Sciences Ciglitazone (25mg). CAS: 74772-77-3
SDP
Supplier:  Enzo Life Sciences BMLGR2050025
Encompass_Preferred
Ciglitazone is a thiazolidinedione type of antihyperglycemic agent. It displays antihyperglycemic activity in genetically obese C57 Bl/6 ob/ob mice and is a selective PPARγ agonist (EC50=3 µM). Inhibits HUVEC differentiation and angiogenesis and also stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0929