Learn More
Medchemexpress LLC Cilazapril monohydrate | 92077-78-6 | 99.9% | C22H33N3O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA0043A10MM/1ML
Encompass_Preferred
Cilazapril monohydrate is an orally active prodrug of the angiotensin-converting enzyme (ACE) inhibitor Cilazaprilat. It reduces plasma ACE activity and can be used in the research of hypertension (including essential and renal hypertension) and congestive heart failure. This product is for research use only and not sold to patients.
  • Orally active prodrug
  • Reduces plasma ACE activity
  • Used in research of hypertension
  • Used in research of congestive heart failure
  • Evokes significant plasma ACE inhibition
  • Reduces blood pressure in dogs with renal hypertension
  • Improves postischemic myocardial function in hyperthyroid rats
Catalog No. 50-004-32031