Learn More
Medchemexpress LLC Cilazapril monohydrate | 92077-78-6 | 99.9% | C22H33N3O6 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA0043A50MG
Encompass_Preferred
Cilazapril monohydrate is an orally active prodrug of an angiotensin-converting enzyme (ACE) inhibitor. It functions by reducing plasma ACE activity and is utilized in research concerning hypertension, including essential and renal hypertension, and congestive heart failure.
  • Used in research of hypertension (including essential and renal hypertension) and congestive heart failure.
  • Evokes maximum degree of plasma ACE inhibition in rats.
  • Dose-dependently reduces plasma ACE activity and pressor response to angiotensin in cats.
  • Gradually reduces blood pressure in dogs with volume-depleted renal hypertension.
  • Decreases blood pressure, reduces heart weight to body weight ratio, and improves postischemic myocardial function in hyperthyroid rats.
Catalog No. 50-004-32032