Learn More
Selleck Chemical LLC Cilengitidetrifluoroacetate 5mg 199807-35-7 EMD 121974, NSC 707544
SDP
Supplier:  Selleck Chemical LLC S70775MG
Encompass_Preferred
Cilengitide (EMD 121974, NSC 707544) is a potent integrin inhibitor for ?v?3 receptor and ?v?5 receptor with IC50 of 4.1 nM and 79 nM in cell-free assays, respectively; ~10-fold selectivity against gpIIbIIIa. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7964