Learn More
Medchemexpress LLC Ciliobrevin A | 302803-72-1 | 98.6% | 358.18 g·mol⁻1 | C17H9Cl2N3O2 | 1 ML

Supplier: Medchemexpress LLC HY10079010MM 1ML
Ciliobrevin A is a hedgehog (Hh) signaling pathway inhibitor supplied as a 10 mM solution in DMSO for research use. It shows reported potency with an IC50 less than 10 μM and has molecular formula C17H9Cl2N3O2 (MW 358.18 g·mol⁻1). Follow recommended handling: aliquot to avoid freeze-thaw, observe storage conditions, and prepare fresh working solutions for in vivo use.
- Provided as a 10 mM solution in DMSO.
- Potent hedgehog pathway inhibitor (IC50 < 10 μM).
- Molecular formula C17H9Cl2N3O2; molecular weight 358.18 g·mol⁻1.
- For research use only; not for human therapeutic use.
- Aliquot to prevent repeated freeze-thaw cycles.
- Recommended storage: -80°C (2 years) or -20°C (1 year).
- Prepare fresh working solutions for in vivo experiments; keep DMSO < 2% in dosing solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.