Learn More
Medchemexpress LLC Cilnidipine | 132203-70-4 | 99.89% | 492.52 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1740410MM/1ML
Encompass_Preferred
Cilnidipine is a long-acting, second-generation dihydropyridine Ca2+-channel blocker that targets L and N-type Ca2+ channels, exhibiting antihypertensive effects. It is suitable for various laboratory applications.
  • Long-acting
  • Second-generation dihydropyridine Ca2+-channel blocker
  • Blocks L and N-type Ca2+ channels
  • Antihypertensive effects
Catalog No. 50-003-85965