Learn More
Apexbio Technology LLC Cilnidipine 132203-70-4 25mg
SDP
Supplier:  Apexbio Technology LLC A144725
Encompass_Preferred
Cilnidipine (CAS 132203-70-4) is a small molecule calcium channel blocker that inhibits both L-type and N-type voltage-gated calcium channels Through this dual antagonism cilnidipine exerts antihypertensive effects and modulates sympathetic neural activity In preclinical studies cilnidipine has been shown to lower mean arterial blood pressure and attenuate cerebral infarct size in rat models of focal brain ischemia indicating neuroprotective properties This compound is utilized in research investigating mechanisms of hypertension neuroprotection and cerebral ischemic injury
Catalog No. 50-224-0732