Learn More
Medchemexpress LLC Cilobradine hydrochloride | 186097-54-1 | MFCD07776908 | 99.7% | 519.07 | C28H39ClN2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18940A10MG
Encompass_Preferred
Cilobradine hydrochloride is a research reagent that blocks hyperpolarization-activated cyclic nucleotide-gated (HCN) channels. It produces a use-dependent blockade of cardiac pacemaker current and modulates HCN1-HCN4 currents in cellular assays, with reported antidepressant-like effects in animal models. The compound is supplied as the hydrochloride salt for preclinical electrophysiology and neuropharmacology studies.
  • HCN channel blocker active on HCN1-HCN4
  • Use-dependent blockade of cardiac pacemaker current
  • Demonstrated antidepressant-like effects in animal models
  • Supplied as hydrochloride salt suitable for research
  • Characterized by CAS 186097-54-1 and molecular weight 519.07
  • High purity (about 99.7%) for experimental use
Catalog No. 50-002-94697