Learn More
Medchemexpress LLC HY-109083 10mg Medchemexpress, Cilofexor CAS:1418274-28-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10908310MG
Encompass_Preferred
Medchemexpress, HY-109083 10mg Cilofexor CAS:1418274-28-8 Cilofexor (GS-9674) is a farnesoid X receptor (FXR) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8667