Learn More
TARGETMOL CHEMICALS INC Cilofexor 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T74365MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cilofexor inhibits binding of a synthetic peptide, Cilofexor is a farnesoid X receptor (FXR) agonist. Purity 98%
Catalog No. 50-287-6830