Learn More
TARGETMOL CHEMICALS INC Cilomilast 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T644510MG
Encompass_Preferred
Cilomilast is a potent PDE4 inhibitor with IC50 of about 110 nM, has anti-inflammatory activity and low central nervous system activity. Phase 3.
Catalog No. NC1624015