Learn More
Selleck Chemical LLC Cilomilast 5mg 153259-65-5 SB-207499
SDP
Supplier:  Selleck Chemical LLC S14555MG
Encompass_Preferred
Cilomilast (SB-207499) is a potent PDE4 inhibitor with IC50 of about 110 nM, has anti-inflammatory activity and low central nervous system activity. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1273