Learn More
Abcam Cilostamide, PDE3 inhibitor, 10MG
Supplier:  Abcam AB14127310MG
Encompass_Preferred
MW 342.4 Da, Purity >98%. Potent, selective PDE3 inhibitor (IC₅₀ values are 27 and 50 nM for PDE3A and PDE3B, respectively). Inhibits ADP-induced platelet aggregation (IC₅₀ = 16.8 μM). Antithrombotic effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6729