Learn More
Bia Separartions Inc CIMac PrimaS - 0.1 Analytical Column (2um)
Supplier:  Bia Separartions Inc 110.51182
Encompass
Multimodal chromatography ligand that combines elements of hydrogen bonding with anion exchange chromatography. Offers unique selectivities in many important product areas. Supports purification of single-stranded RNA at ambient temperature and also fractionates ssRNA by size.
Catalog No. NC1862237