Learn More
Selleck Chemical LLC Cimetidine S1845-50mg
SDP
Supplier:  Selleck Chemical LLC S184550MG
Encompass_Preferred
Cimetidine (SKF-92334) a histamine congener competitively inhibits histamine binding to histamine H2 receptors
Catalog No. 50-310-9590