Learn More
Selleck Chemical LLC Cimetidine S1845-1g
SDP
Supplier:  Selleck Chemical LLC S18451G
Encompass_Preferred
Cimetidine (SKF-92334) a histamine congener competitively inhibits histamine binding to histamine H2 receptors
Catalog No. 50-311-0598