Learn More
Medchemexpress LLC HY-U00106 1mg Medchemexpress, Cimetropium (Bromide) CAS:51598-60-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001061MG
Encompass_Preferred
Medchemexpress, HY-U00106 1mg Cimetropium (Bromide) CAS:51598-60-8 Cimetropium Bromide (DA-3177) is a mAChR antagonist for long-term treatment of irritable bowel syndrome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4416