Learn More
Cayman Chemical CInabarInIc AcId 10mg
SDP
Supplier:  Cayman Chemical 1198810 MG
Encompass_Preferred
An agonist of mGluR4 and active metabolite of 3-hydroxyanthranilic acid; selectively induces inositol phosphate production in HEK293 cells expressing mGluR4 over HEK293 cells expressing mGluR1, mGluR2, mGluR5, mGluR6, mGluR7, or mGluR8 at 100 µM; inhibits DAAO in a cell-free assay (IC50 = 1.37 µM); increases levels of ROS and induces apoptosis in primary mouse thymocytes at 30 µM
Catalog No. 50-295-5786