Learn More
Cayman Chemical CInabarInIc AcId-d4 5mg
SDP
Supplier:  Cayman Chemical 372095 MG
Encompass_Preferred
An internal standard for the quantification of cinnabarinic acid by GC- or LC-MS
Catalog No. 50-291-4842