Learn More
TARGETMOL CHEMICALS INC CINACALCET 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T8403100MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cinacalcet (AMG 073) is an orally active allosteric Ca receptor (CaR) agonist treatment of cardiovascular disease . purity: 99%
Catalog No. 50-257-9079