Learn More
Medchemexpress LLC Cinacalcet | 226256-56-0 | 99.9% | 357.41 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY70037
Encompass_Preferred
Cinacalcet is an orally active, allosteric agonist of the Ca receptor (CaR), primarily used for cardiovascular disease research. In in vivo studies, it has been shown to significantly reduce parathyroid gland weight in 5/6 nephrectomy animals and blood ionized calcium levels in sham animals. It also causes a marked reduction in circulating parathyroid hormone.
  • Orally active Ca receptor agonist
  • Reduces parathyroid gland weight in nephrectomy models
  • Decreases blood ionized calcium levels
  • Lowers circulating parathyroid hormone
  • White to off-white solid appearance
  • Stable for extended periods when stored as powder or in solvent
Catalog No. 50-003-64803