Learn More
Medchemexpress LLC Cinacalcet impurity 1 hydrochloride | 1229225-42-6 | MFCD28167915 | >98.0% | 365.82 | C20H19ClF3N | 10 MG
SDP
Supplier:  Medchemexpress LLC HYI0577A10MG
Encompass_Preferred
Cinacalcet impurity 1 hydrochloride is a characterized impurity of the calcimimetic cinacalcet, provided as a research reference standard for analytical method development, impurity profiling, and related laboratory research. It is intended for research use only and not for human or clinical applications.
  • High chemical purity suitable for analytical reference and quality control.
  • Available in small-scale quantities for method development and validation.
  • Supplied with CAS, formula, and molecular weight for unambiguous identification.
  • Stored and shipped under recommended conditions to maintain stability.
  • Intended for use as a drug intermediate or impurity reference standard.
Catalog No. 50-002-96970