Learn More
Medchemexpress LLC Cinacalcet impurity 9 | 253337-60-9 | 303.45 | C22H25N | 10 MG

Supplier: Medchemexpress LLC HYI019210MG
Cinacalcet impurity 9 is a characterized impurity of cinacalcet supplied as a reference standard for analytical and research applications. The material is provided as a weighed solid in small mass quantities for impurity profiling and analytical method development.
- Intended for analytical reference and impurity testing.
- CAS number 253337-60-9.
- Molecular formula C22H25N; molecular weight 303.45.
- Available in small mass packaging suitable for method development.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.