Learn More
Medchemexpress LLC Cinacase impurity 12 | 1229224-93-4 | 98.5% | 361.44 | C22H26F3N | 10 MG

Supplier: Medchemexpress LLC HYI043710MG
Cinacase impurity 12 is an identified impurity of cinacase provided for laboratory research and analytical use. It is supplied as a high-purity liquid suitable as an analytical reference and for stability testing.
- Purity 98.48%
- CAS number 1229224-93-4
- Molecular weight 361.44
- Chemical formula C22H26F3N
- Appearance liquid
- Storage: pure form -20°C (3 years) or 4°C (2 years)
- In solvent: -80°C (6 months) or -20°C (1 month)
- Available sizes include 5 mg, 10 mg, 25 mg, 50 mg, 100 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.