Learn More
Medchemexpress LLC Cinaciguat hydrochloride | 646995-35-9 | 98.9% | C36H40ClNO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14181A100MG
Encompass_Preferred
Cinaciguat hydrochloride is a potent soluble guanylate cyclase (GC) activator. It exhibits an EC50 of 15 nM in platelets, making it a valuable tool for studying guanylate cyclase pathways and related biological activities.
  • Potent guanylate cyclase activator
  • Effective at nanomolar concentrations in platelets
  • Store solid at 4°C in a sealed container, away from moisture
  • Store solutions at -80°C for up to 6 months or -20°C for up to 1 month, sealed and away from moisture
Catalog No. 50-003-56544