Learn More
Medchemexpress LLC Cinakase impurity 1 | 1229224-94-5 | 95.3% | 373.41 g/mol | C22H22F3NO | 10 MG

Supplier: Medchemexpress LLC HYI019410MG
Cinakase impurity 1 is an isolated impurity of the investigational compound Cinakase provided for research and analytical applications. It is intended as an analytical reference and synthesis intermediate for impurity profiling, method development, and analytical validation.
- Reported purity 95.3%.
- Molecular formula C22H22F3NO.
- Molecular weight 373.41 g/mol.
- CAS number 1229224-94-5.
- Available in small-scale packages for analytical use, e.g., 10 MG.
- Store under recommended conditions; long-term storage -20°C under nitrogen.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.