Learn More
Medchemexpress LLC Cinakase impurity 13 | 2074615-22-6 | 98.8% | 357.41 | C22H22F3N | 10 MG

Supplier: Medchemexpress LLC HYI039110MG
Cinakase impurity 13 is a chemical impurity related to the investigational compound Cinakase, supplied for research use and impurity characterization. It is suitable for analytical method development, reference standard work, and compound profiling.
- Chemical formula C22H22F3N.
- Molecular weight 357.41 g/mol.
- CAS number 2074615-22-6.
- Purity approximately 98.8%.
- Available in small mg quantities suitable for analytical use, including 10 MG vials.
- Store under conditions specified in the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.