Learn More
Cayman Chemical CInamtnIn B-1 10mg
SDP
Supplier:  Cayman Chemical 2169410 MG
Encompass_Preferred
A polyphenol antioxidant; inhibits lipid peroxidation in vitro (IC50 = 2.25 µM); improves wound healing in mice; COX-2 inhibitor with 19, 27, and 86% inhibition in Sf9 cells at 10, 100, and 1,000 UG/ml, respectively
Catalog No. 50-298-6308